Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse RPTOR Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC5957N100UG
This item is not returnable.
View return policy
Description
RPTOR Monoclonal specifically detects RPTOR in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
RPTOR | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
KIAA1303, KIAA1303p150 target of rapamycin (TOR)-scaffold protein containing WD-repeats, KOG1, Mip1, p150 target of rapamycin (TOR)-scaffold protein, Raptor, regulatory associated protein of MTOR, complex 1, regulatory-associated protein of mTOR | |
E. coli-derived recombinant human RPTOR Lys77-Gln230 Accession # Q8N122 | |
RUO | |
57521 | |
Human, Mouse | |
IgG2b |
Flow Cytometry | |
514208 | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q8N122 | |
Mouse | |
100 μg | |
Primary | |
Detects human and mouse RPTOR in Western blots. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction