Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse RUNX3/CBFA3 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC3765G100UG
This item is not returnable.
View return policy
Description
RUNX3/CBFA3 Monoclonal specifically detects RUNX3/CBFA3 in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
RUNX3/CBFA3 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q13761 | |
RUNX3 | |
E. coli-derived recombinant human RUNX3/CBFA3 Lys186-Tyr415 Accession # Q13761 | |
RUO | |
864 | |
Human, Mouse | |
IgG2a |
Flow Cytometry | |
527327 | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Acute myeloid leukemia 2 protein, AML2, AML2SL3/AKV core-binding factor alpha C subunit, CBFA3, CBFA3MGC16070, CBF-alpha-3, PEA2 alpha C, PEA2-alpha C, PEBP2 alpha C, PEBP2A3, PEBP2A3FLJ34510, PEBP2AC, PEBP2-alpha C, runt domain, alpha subunit 3, runt-related transcription factor 3, SL3-3 enhancer factor 1 alpha C subunit, transcription factor AML2 | |
Mouse | |
100 μg | |
Primary | |
Detects human and mouse RUNX3/CBFA3 in Western blots. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction