Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse S100A6 Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF4584
This item is not returnable.
View return policy
Description
S100A6 Polyclonal specifically detects S100A6 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Knockout Validated.Specifications
S100A6 | |
Polyclonal | |
Unconjugated | |
P14069 | |
S100A6 | |
E. coli-derived recombinant mouse S100A6 Met1-Lys89 Accession # P14069 | |
100 μg | |
Primary | |
Detects human and mouse S100A6 in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant mouse S100A1 and recombinant mouse S100A4 is observed. | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunoassay | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL, Knockout Validated | |
2A9, CABP, CACY, CACY5B10, calcyclin, Growth factor-inducible protein 2A9, MLN 4, PRA, PRAS100 calcium binding protein A6 (calcyclin), Prolactin receptor-associated protein, protein S100-A6, S100 calcium binding protein A6, S100 calcium-binding protein A6, S100 calcium-binding protein A6 (calcyclin) | |
Sheep | |
Affinity Purified | |
RUO | |
6277 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction