Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse S100A6 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF4584
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF4584 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF4584 Supplier R&D Systems Supplier No. BAF4584
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

S100A6 Polyclonal specifically detects S100A6 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen S100A6
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. P14069
Gene Alias 2A9, CABP, CACY, CACY5B10, calcyclin, Growth factor-inducible protein 2A9, MLN 4, PRA, PRAS100 calcium binding protein A6 (calcyclin), Prolactin receptor-associated protein, protein S100-A6, S100 calcium binding protein A6, S100 calcium-binding protein A6, S100 calcium-binding protein A6 (calcyclin)
Gene Symbols S100A6
Host Species Sheep
Immunogen E. coli-derived recombinant mouse S100A6 Met1-Lys89 Accession # P14069
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6277
Test Specificity Detects human and mouse S100A6 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human (rh) S100A1, rhS100A2, and recombinant mouse S100A4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.