Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse SALL4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6374
This item is not returnable.
View return policy
Description
SALL4 Monoclonal specifically detects SALL4 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SALL4 | |
Monoclonal | |
LYOPH | |
Western Blot 0.025 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Q9UJQ4 | |
SALL4 | |
E. coli-derived recombinant human SALL4 Lys96-Gly359 Accession # Q9UJQ4 | |
100 μg | |
Primary | |
Detects endogenous human and mouse SALL4 in Western blots. | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry | |
651839 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
dJ1112F19.1, DRRS, HSAL4, MGC133050, sal-like 4 (Drosophila), sal-like protein 4, Zinc finger protein 797, Zinc finger protein SALL4, ZNF797, ZNF797sal (Drosophila)-like 4 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
57167 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction