Learn More
Human/Mouse Semaphorin 3A Alexa Fluor 488-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | Semaphorin 3A |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 215803 |
| Conjugate | Alexa Fluor 488 |
| Dilution | Intracellular Staining by Flow Cytometry 5 uL/10^6 cells |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide |
| Gene Accession No. | Q14563 |
| Gene Alias | (semaphorin) 3A, coll-1, collapsin 1, Hsema-I, Hsema-III, MGC133243, sema domain, immunoglobulin domain (Ig), short basic domain, secreted, Sema III, SEMA1, Sema3A, SEMAD, SEMAIII, SEMAL, semaphorin D, Semaphorin III, semaphorin L, semaphorin-3A, SemD |
| Gene Symbols | SEMA3A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.