Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Serpin I1 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $491.00
Specifications
Antigen | Serpin I1/Neuroserpin |
---|---|
Clone | 437016 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Serpin I1/Neuroserpin Monoclonal specifically detects Serpin I1/Neuroserpin in Human, Mouse samples. It is validated for Western Blot.Specifications
Serpin I1/Neuroserpin | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Human, Mouse | |
AAH18043 | |
5274 | |
Mouse myeloma cell line NS0-derived recombinant human Serpin I1 Ala19-Leu410 Accession # AAH18043 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human and mouse SerpinI1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross reactivity with recombinant mouse Serpin I1 is observed and no cross-reactivity with recombinant human Serpin A1, A3, A4, A5, A12, B6, B8, B9, C1, D1, F2, G1, or I2 is observed. |
437016 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DKFZp781N13156, Neuroserpin, Peptidase inhibitor 12, PI12, PI-12, PI12Serpin I1, serine (or cysteine) proteinase inhibitor, clade I (neuroserpin), member 1, serpin peptidase inhibitor, clade I (neuroserpin), member 1 | |
SERPINI1 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title