Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Serum Albumin Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3329
This item is not returnable.
View return policy
Description
Albumin Polyclonal specifically detects Albumin in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.Specifications
Albumin | |
Polyclonal | |
Western Blot 0.1 ug/mL, Simple Western 5-10 ug/mL, Immunocytochemistry 5-15 ug/mL | |
ALB, albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin | |
Goat | |
Affinity Purified | |
RUO | |
213 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ALB | |
Mouse Serum Albumin | |
100 μg | |
Primary | |
Detects human and mouse Serum Albumin in direct ELISAs and Western blots. | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction