Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse SEZ6L2/BSRP-A Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF4916
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF4916 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF4916 Supplier R&D Systems Supplier No. BAF4916
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

SEZ6L2/BSRP-A Polyclonal specifically detects SEZ6L2/BSRP-A in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen SEZ6L2/BSRP-A
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q4V9Z5
Gene Alias BSRPA, BSRP-A, PSK-1, seizure 6-like protein 2, seizure related 6 homolog (mouse)-like 2, UNQ1903/PRO4349
Gene Symbols SEZ6L2
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse SEZ6L2/BSRP-A Leu28-Ser837 Accession # Q4V9Z5
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26470
Test Specificity Detects human and mouse SEZ6L2/BSRP-A in Western blots. In Western blots, less than 5% cross-reactivity with recombinant mouse (rm) BSRP-B and rmBSRP-C is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.