Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse sFRP-3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1921SP
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1921SP 25 μg
MAB1921 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1921SP Supplier R&D Systems Supplier No. MAB1921SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

sFRP-3/FRZB Monoclonal specifically detects sFRP-3/FRZB in Human, Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen sFRP-3/FRZB
Applications Western Blot, ELISA
Classification Monoclonal
Clone 133931
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAB51298
Gene Alias FIZ, FREOS1, Frezzled, Fritz, frizzled-related protein, Frizzled-related protein 1, FRP, FRP-3, FRZB, Frzb1, FrzB-1, FRZB1sFRP-3, FRZB-PEN, FZRB, hFIZ, secreted frizzled-related protein 3, sFRP3, SFRP3frezzled, SRFP3frizzled homolog-related
Gene Symbols FRZB
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human sFRP-3 Ala33-Asn325 Accession # AAB51298
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2487
Test Specificity Detects human and mouse sFRP-3 in ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.