Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse sFRP-3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB192050
Change view
Click to view available options
Quantity:
25 μg
50 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB192050 50 μg
MAB192SP 25 μg
MAB192500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB192050 Supplier R&D Systems Supplier No. MAB192050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

sFRP-3/FRZB Monoclonal specifically detects sFRP-3/FRZB in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen sFRP-3/FRZB
Applications Western Blot
Classification Monoclonal
Clone 133912
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAB51298
Gene Alias FIZ, FREOS1, Frezzled, Fritz, frizzled-related protein, Frizzled-related protein 1, FRP, FRP-3, FRZB, Frzb1, FrzB-1, FRZB1sFRP-3, FRZB-PEN, FZRB, hFIZ, secreted frizzled-related protein 3, sFRP3, SFRP3frezzled, SRFP3frizzled homolog-related
Gene Symbols FRZB
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human sFRP-3 Ala33-Asn325 Accession # AAB51298
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2487
Test Specificity Detects human sFRP-3 in direct ELISAs and Western blots. Shows 100% cross-reactivity with recombinant mouse (rm) sFRP-3 and no cross-reactivity with recombinant human (rh) sFRP-1, rmsFRP-2, or rhsFRP-4.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.