Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse SHIP Antibody, R&D Systems™

Goat Antibody
Supplier: R&D Systems AF10117100
This item is not returnable.
View return policy
Description
SHIP specifically detects SHIP in Human, Mouse samples. It is validated for Western Blot.Specifications
SHIP | |
Polyclonal | |
Western Blot 1 μ/mL | |
Q9ES52 | |
Goat | |
100 μg | |
3635 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Western Blot | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
145kD, EC 3.1.3, hp51CNMGC142142, Inositol polyphosphate-5-phosphatase of 145 kDa, inositol polyphosphate-5-phosphatase, 145kDa, INPP5D, MGC104855, SHIPMGC142140 | |
E. coli-derived recombinant mouse SHIP, Glu874-Thr941, Accession No. Q9ES52 | |
Primary | |
Detects mouse SHIP in direct ELISAs. | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction