Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Slit2 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB5444U100UG
This item is not returnable.
View return policy
Description
Slit2 Monoclonal specifically detects Slit2 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
Slit2 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9R1B9 | |
SLIT2 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse Slit2 Gln26-Gln900 Accession # Q9R1B9 | |
RUO | |
9353 | |
Human, Mouse | |
IgG2b |
Flow Cytometry | |
710305 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
FLJ14420, SLIL3, slit (Drosophila) homolog 2, slit homolog 2 (Drosophila), slit homolog 2 protein, Slit-2SLIL3 | |
Rat | |
100 μg | |
Primary | |
Stains human and mouse Slit2 transfectants but not irrelevant transfectants in flow cytometry. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction