Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Slit2 Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB5444V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB5444V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB5444V100 Supplier R&D Systems Supplier No. FAB5444V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Slit2 Monoclonal specifically detects Slit2 in Human, Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Slit2
Applications Flow Cytometry
Classification Monoclonal
Clone 710305
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q9R1B9
Gene Alias FLJ14420, SLIL3, slit (Drosophila) homolog 2, slit homolog 2 (Drosophila), slit homolog 2 protein, Slit-2SLIL3
Gene Symbols SLIT2
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Slit2 Gln26-Gln900 Accession # Q9R1B9
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9353
Test Specificity Stains human and mouse Slit2 transfectants but not irrelevant transfectants in flow cytometry.
Target Species Human, Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.