Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Smad9 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7215
This item is not returnable.
View return policy
Description
Smad9 Polyclonal specifically detects Smad9 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry.Specifications
Smad9 | |
Polyclonal | |
Unconjugated | |
O15198 | |
SMAD9 | |
E. coli-derived recombinant human Smad9 Ala158-Ser223 Accession # O15198 | |
100 μg | |
Primary | |
Detects human Smad9 in direct ELISAs. In direct ELISAs, approximately 5% cross-reactivity with recombinant human Smad1 is observed. | |
Human, Mouse | |
IgG |
Immunohistochemistry, Immunocytochemistry | |
LYOPH | |
Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL | |
MAD homolog 9, MAD, mothers against decapentaplegic homolog 9 (Drosophila), Madh6, MADH6mothers against decapentaplegic homolog 9, MADH9, MADH9Mothers against decapentaplegic, drosophila, homolog of, 9, Mothers against DPP homolog 9, SMAD 9, SMAD family member 9SMAD8B, SMAD, mothers against DPP homolog 9, SMAD, mothers against DPP homolog 9 (Drosophila), SMAD8, SMAD8A, Smad9 | |
Sheep | |
Affinity Purified | |
RUO | |
4093 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction