Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse SOD1/Cu-Zn SOD Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF3418SP
This item is not returnable.
View return policy
Description
SOD1/Cu-Zn SOD Polyclonal specifically detects SOD1/Cu-Zn SOD in Human, Mouse samples. It is validated for Western Blot, Simple Western.Specifications
SOD1/Cu-Zn SOD | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ALS, ALS1, amyotrophic lateral sclerosis 1 (adult), Cu, Cu/Zn superoxide dismutase, CuZn SOD, Cu-Zn SOD, EC 1.15.1.1, homodimer, hSod1, indophenoloxidase A, Ipo1, IPOA, SOD, SOD, cytosolic, SOD, Soluble, superoxide dismutase [Cu-Zn], Superoxide dismutase 1, superoxide dismutase 1, soluble, Zn superoxide dismutase, EC 1.15.1.110superoxide dismutase, cystolic | |
Goat | |
Affinity Purified | |
RUO | |
6647 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.2 ug/mL, Simple Western 10 ug/mL | |
P00441 | |
SOD1 | |
E. coli-derived recombinant human SOD1/Cu-Zn SOD Met1-Gln154 Accession # P00441 | |
25 μg | |
Primary | |
Detects human and mouse SOD1/Cu-Zn SOD in Western blots. Detects rat SOD1, but Catalog # AF3787 is recommended. In Western blots, less than 1% cross-reactivity with recombinant human SOD2 or SOD3 is observed. | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction