Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse SOD1/Cu-Zn SOD Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF3418
This item is not returnable.
View return policy
Description
SOD1/Cu-Zn SOD Polyclonal specifically detects SOD1/Cu-Zn SOD in Human, Mouse samples. It is validated for Western Blot, Simple Western.Specifications
SOD1/Cu-Zn SOD | |
Polyclonal | |
Unconjugated | |
P00441 | |
SOD1 | |
E. coli-derived recombinant human SOD1/Cu-Zn SOD Met1-Gln154 Accession # P00441 | |
50 μg | |
Primary | |
Detects human and mouse SOD1/Cu-Zn SOD in Western blots. Detects rat SOD1, but Catalog # AF3787 is recommended. In Western blots, less than 1% cross-reactivity with recombinant human SOD2 or SOD3 is observed. | |
Human, Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.2 ug/mL, Simple Western 10 ug/mL | |
ALS, ALS1, amyotrophic lateral sclerosis 1 (adult), Cu, Cu/Zn superoxide dismutase, CuZn SOD, Cu-Zn SOD, EC 1.15.1.1, homodimer, hSod1, indophenoloxidase A, Ipo1, IPOA, SOD, SOD, cytosolic, SOD, Soluble, superoxide dismutase [Cu-Zn], Superoxide dismutase 1, superoxide dismutase 1, soluble, Zn superoxide dismutase, EC 1.15.1.110superoxide dismutase, cystolic | |
Goat | |
Affinity Purified | |
RUO | |
6647 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction