Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Somatostatin Antibody, R&D Systems™

Rat Monoclonal Antibody
$227.50 - $573.00
Specifications
Antigen | Somatostatin |
---|---|
Clone | 906552 |
Dilution | Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL |
Applications | Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Description
Somatostatin Monoclonal specifically detects Somatostatin in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry.Specifications
Somatostatin | |
Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Growth hormone release-inhibiting factor, SMST, SOM, somatostatin, somatostatin-14, somatostatin-28, SRIF, SST | |
SST | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
906552 | |
Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Rat | |
Human, Mouse | |
P61278 | |
6750 | |
KLH-coupled human Somatostatin peptide Accession # P61278 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Somatostatin in direct ELISA. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title