Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Sonic Hedgehog/Shh N-Terminus PE-conjugated Antibody, R&D Systems™

Catalog No. IC4641P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC4641P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC4641P Supplier R&D Systems Supplier No. IC4641P
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 2 publications

Sonic Hedgehog/Shh Monoclonal specifically detects Sonic Hedgehog/Shh in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen Sonic Hedgehog/Shh
Applications Flow Cytometry
Classification Monoclonal
Clone 171018
Conjugate R-PE
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q62226
Gene Alias HHG1, HHG-1, HLP3, HPE3, MCOPCB5, MCOPCB5sonic hedgehog (Drosophila) homolog, Shh, ShhNC, SMMCI, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS
Gene Symbols SHH
Host Species Rat
Immunogen E. coli-derived recombinant mouse Sonic Hedgehog/SHH N-terminus Cys25-Gly198 (Lys122Arg) Accession # Q62226
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6469
Test Specificity Detects human and mouse Sonic Hedgehog/SHH in Western blots and mouse Sonic Hedgehog/SHH in ELISAs. In Western blots, approximately 50% cross-reactivity with recombinant mouse (rm) Desert Hedgehog/Dhh (N-terminus; aa 23-198) is observed and no cross-reactivity with rmIndian Hedgehog/Ihh (N-terminus; aa 66-240), rmDesert Hedgehog/Dhh (C-terminus; aa 199-396), or rmSonic Hedgehog/Shh (C-terminus; aa 199-437) is observed.
Target Species Human, Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.