Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse SPRY2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6157SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6157SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6157SP Supplier R&D Systems Supplier No. AF6157SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

SPRY2 Polyclonal specifically detects SPRY2 in Human, Mouse samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen SPRY2
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 50 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O43597
Gene Alias hSPRY2, MGC23039, protein sprouty homolog 2, sprouty (Drosophila) homolog 2, sprouty 2, sprouty homolog 2 (Drosophila), spry-2
Gene Symbols SPRY2
Host Species Goat
Immunogen E. coli-derived recombinant human SPRY2 Met1-Ala175 Accession # O43597
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10253
Test Specificity Detects human and mouse SPRY2 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.