Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse SR140 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB9897100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9897-100 100 μg
MAB9897-SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9897-100 Supplier R&D Systems Supplier No. MAB9897100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SR140 Monoclonal specifically detects SR140 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen SR140
Applications Western Blot
Classification Monoclonal
Clone 983615
Conjugate Unconjugated
Dilution Western Blot 2μg/mL
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Gene Accession No. O15042
Gene Alias 140 kDa Ser/Arg-rich domain protein, fSAPa, KIAA0332, SR140, U2 snRNP-associated SURP domain containing, U2 snRNP-associated SURP motif-containing protein, U2-associated protein SR140, U2-Associated SR140 Protein, U2SURP
Host Species Mouse
Immunogen Synthetic peptide containing human SR140 Peptide Accession # O15042
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 23350
Test Specificity Detects human SR140 in direct ELISAs. Detects human and mouse SR140 in Western blots.
Reconstitution Reconstitute at 0.5mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.