Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse SSEA-3 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB1434G
This item is not returnable.
View return policy
Description
SSEA-3 Monoclonal specifically detects SSEA-3 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
SSEA-3 | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
Rat | |
Affinity Purified | |
RUO | |
Recognizes a carbohydrate epitope of SSEA-3 (4, 5). | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Flow Cytometry | |
MC-631 | |
Flow Cytometry 5 uL/10^6 cells | |
SSEA3, Stage specific embryonic antigen 3, Stage-specific embryonic antigen 3 | |
Four to eight cell stage mouse embryos | |
100 Tests | |
Primary | |
Human, Mouse | |
IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction