Learn More
Human/Mouse SSEA-4 Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 38 publications
Supplier:  R&D Systems MAB1435
Catalog No. MAB1435
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
SSEA-4 Monoclonal specifically detects SSEA-4 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry.
Specifications
Specifications
SSEA-4
Monoclonal
LYOPH
Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Recognizes a carbohydrate epitope of SSEA-4 (1, 2).
Human, Mouse
Purified
Flow Cytometry, Immunocytochemistry
MC-813-70
Unconjugated
Lyophilized from a 0.2 μm filtered solution in MES with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
2120Ep human embryonal carcinoma cell line
100 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG3
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.