Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse SSEA-4 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1435P025
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1435P025 25 Tests
FAB1435P100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB1435P025 Supplier R&D Systems Supplier No. FAB1435P025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 14 publications

SSEA-4 Monoclonal specifically detects SSEA-4 in Human, Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen SSEA-4
Applications Flow Cytometry
Classification Monoclonal
Clone MC-813-70
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Host Species Mouse
Immunogen 2120Ep human embryonal carcinoma cell line
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes a carbohydrate epitope of SSEA-4 (1, 2).
Target Species Human, Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.