Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse STAT5a Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2174
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2174 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB2174 Supplier R&D Systems Supplier No. MAB2174
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

STAT5A Monoclonal specifically detects STAT5A in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen STAT5a
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 251619
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_003143
Gene Alias MGF, signal transducer and activator of transcription 5A, STAT5
Gene Symbols STAT5A
Host Species Mouse
Immunogen Human STAT5a synthetic peptide SLDSRLSPPAGLFTSARGSLS Accession # NP_003143
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6776
Test Specificity Detects human and mouse STAT5a. Does not cross-react with STAT5b.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.