Learn More
Human/Mouse STAT5b Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | STAT5b |
| Applications | Western Blot, Immunoprecipitation, Flow Cytometry, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.2 ug/mL, Simple Western 10 ug/mL, Immunoprecipitation 1-5 ug/500 ug cell lysate, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready, Knockout Validated |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Alias | signal transducer and activator of transcription 5B, STAT5, transcription factor STAT5B |
| Gene Symbols | STAT5B |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.