Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Stathmin-2/STMN2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6930
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6930 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6930 Supplier R&D Systems Supplier No. MAB6930
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Stathmin-2/STMN2 Monoclonal specifically detects Stathmin-2/STMN2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Stathmin-2/STMN2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 684433
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.05 - 0.2 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q93045
Gene Alias FLJ34868, FLJ50995, neuron-specific growth-associated protein, Protein SCG10, SCG10, SCG10neuronal growth-associated protein (silencer element), SCGN10, SCGN10stathmin-2, Stathmin2, stathmin-like 2, STMN2, superior cervical ganglia, neural specific 10, Superior cervical ganglion-10 protein
Gene Symbols STMN2
Host Species Mouse
Immunogen E. coli-derived recombinant human Stathmin-2/STMN2 Tyr26-Gly179 Accession # Q93045
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11075
Test Specificity Detects human Stathmin-2/STMN2 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Stathmin-1, -3, or -4 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.