Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Sulfamidase/SGSH Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB83801100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB83801100 100 μg
MAB83801SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB83801100 Supplier R&D Systems Supplier No. MAB83801100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sulfamidase/SGSH Monoclonal specifically detects Sulfamidase/SGSH in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Sulfamidase/SGSH
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 1018331
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunohistochemistry 5-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P51688
Gene Alias heparan sulfate sulfatase, HSS, HSSN-sulphoglucosamine sulphohydrolase, MPS3A, MPS3Asulphamidase, N-sulfoglucosamine sulfohydrolase, SFMD, SFMDEC 3.10.1.1, SGSH, Sulfoglucosamine sulfamidase, Sulphamidase
Host Species Mouse
Immunogen Spodoptera frugiperda, Sf 21 (baculovirus)-derived human Sulfamidase/SGSH, Arg23-Leu502, Accession # P51688
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6448
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.