Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Sulfatase Modifying Factor 1/SUMF1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2779
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2779 100 μg
MAB2779SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2779 Supplier R&D Systems Supplier No. MAB2779
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sulfatase Modifying Factor 1/SUMF1 Monoclonal specifically detects Sulfatase Modifying Factor 1/SUMF1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Sulfatase Modifying Factor 1/SUMF1
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 329005
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8NBK3.3
Gene Alias AAPA3037, EC 1.8.99.-, FGE1, FGEC-alpha-formylglycine-generating enzyme 1, FGly-generating enzyme, MGC131853, MGC150436, sulfatase modifying factor 1, sulfatase-modifying factor 1, UNQ3037
Gene Symbols SUMF1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Sulfatase Modifying Factor 1/SUMF1 isoform 1 Ser34-Asp374 (Ser63Asn) Accession # Q8NBK3.3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285362
Test Specificity Detects recombinant human or mouse Sulfatase Modifying Factor 1/SUMF1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human SUMF2 or recombinant mouse SUMF2 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.