Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse SUMO1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3289
This item is not returnable.
View return policy
Description
SUMO1 Polyclonal specifically detects SUMO1 in Human, Mouse samples. It is validated for Western Blot.Specifications
SUMO1 | |
Polyclonal | |
Unconjugated | |
P63165 | |
SUMO1 | |
E. coli-derived recombinant human SUMO1 Ser2-Val101 Accession # P63165 | |
100 μg | |
Primary | |
Detects endogenous human and mouse proteins modified with SUMO1 in Western blots. In Western blots, less than 5% cross-reactivity with recombinant human (rh) SUMO2 and rhSUMO3 and less than 1% cross-reactivity with rhSUMO4 is observed. | |
Human, Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
DAP1, GAP modifying protein 1, GAP-modifying protein 1, GMP1SMT3CSMT3H3OFC10UBL1PIC1, PIC1, SENP2, sentrin, small ubiquitin-related modifier 1, SMT3, SMT3 homolog 3, SMT3 suppressor of mif two 3 homolog 1 (S. cerevisiae), SMT3 suppressor of mif two 3 homolog 1 (yeast), Smt3C, SMT3H3, SUMO-1, Ubiquitin-homology domain protein PIC1, ubiquitin-like 1 (sentrin), Ubiquitin-like protein SMT3C, Ubiquitin-like protein UBL1, UBL1 | |
Goat | |
Affinity Purified | |
RUO | |
7341 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction