Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse SUMO2/3/4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3020SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3020SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3020SP Supplier R&D Systems Supplier No. AF3020SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

SUMO2/3/4 Polyclonal specifically detects SUMO2/3/4 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen SUMO2/3/4
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P61956
Host Species Goat
Immunogen E. coli-derived recombinant human SUMO2 Ala2-Tyr95 Accession # P61956
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6613
Test Specificity Detects endogenous human and mouse proteins modified with SUMO2, 3, or 4 in Western blots. Because of the high level of sequence homology, this antibody has equivalent reactivity to SUMO2, SUMO3, and SUMO4 based in Western blots with recombinant SUMO protein. Cross-reactivity to recombinant human SUMO1 was minimal in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.