Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Syntaxin 12 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6617SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6617SP 25 μg
AF6617100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF6617SP Supplier R&D Systems Supplier No. AF6617SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Antibody

Syntaxin 12 specifically detects Syntaxin 12 in Human, Mouse samples. It is validated for Immunohistochemistry.

Specifications

Antigen Syntaxin 12
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1-15 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q86Y82
Gene Alias MGC51957, STX12, STX13, STX14, syntaxin 12, syntaxin-12
Host Species Goat
Immunogen E.coli-derived recombinant human Syntaxin 12, Met1-Gln200, Accession No. Q86Y82
Purification Method Antigen Affinity-purified
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 23673.0
Test Specificity Detects human Syntaxin 12 in direct ELISAs. Detects human and mouse Syntaxin 12 in immunohistochemistry.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.