Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse TBX5 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5918SP
This item is not returnable.
View return policy
Description
TBX5 Polyclonal specifically detects TBX5 in Human, Mouse samples. It is validated for Western Blot, Simple Western.Specifications
TBX5 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
HOS, T-box 5, T-box protein 5, T-box transcription factor TBX5 | |
Sheep | |
Affinity Purified | |
RUO | |
6910 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 50 ug/mL | |
Q99593 | |
TBX5 | |
E. coli-derived recombinant human TBX5 Lys253-Lys327 Accession # Q99593 | |
25 μg | |
Primary | |
Detects human and mouse TBX5 in direct ELISAs and Western blots. | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction