Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse TBX5 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5918
This item is not returnable.
View return policy
Description
TBX5 Polyclonal specifically detects TBX5 in Human, Mouse samples. It is validated for Western Blot, Simple Western.Specifications
TBX5 | |
Polyclonal | |
Unconjugated | |
Q99593 | |
TBX5 | |
E. coli-derived recombinant human TBX5 Lys253-Lys327 Accession # Q99593 | |
100 μg | |
Primary | |
Detects human and mouse TBX5 in direct ELISAs and Western blots. | |
Human, Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 50 ug/mL | |
HOS, T-box 5, T-box protein 5, T-box transcription factor TBX5 | |
Sheep | |
Affinity Purified | |
RUO | |
6910 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction