Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Tec Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6519SP
This item is not returnable.
View return policy
Description
Tec Monoclonal specifically detects Tec in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Tec | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
P42680 | |
TEC | |
E. coli-derived recombinant human Tec Met1-Met187 Accession # P42680 | |
25 μg | |
Primary | |
Detects human Tec in direct ELISAs and Western blots. | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry | |
663921 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 2.7.10, EC 2.7.10.2, MGC126760, MGC126762, PSCTK4, PSCTK4tyrosine-protein kinase Tec, tec protein tyrosine kinase | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7006 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction