Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse TLE3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5785SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5785SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5785SP Supplier R&D Systems Supplier No. AF5785SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

TLE3 Polyclonal specifically detects TLE3 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen TLE3
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q04726
Gene Alias ESG3, ESG3Enhancer of split groucho-like protein 3, ESGenhancer of split groucho 3, GRG3, HsT18976, KIAA1547FLJ39460, transducin-like enhancer of split 3 (E(sp1) homolog, Drosophila), transducin-like enhancer of split 3, homolog of Drosophila E(sp1), transducin-like enhancer protein 3
Gene Symbols TLE3
Host Species Goat
Immunogen E. coli-derived recombinant human TLE3 His11-Asn200 Accession # Q04726
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7090
Test Specificity Detects endogenous human TLE3 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.