Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse TORC2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6338
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6338 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6338 Supplier R&D Systems Supplier No. MAB6338
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TORC2 Monoclonal specifically detects TORC2 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen TORC2
Applications Western Blot
Classification Monoclonal
Clone 628430
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.01 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q53ET0
Gene Alias CREB regulated transcription coactivator 2, CREB-regulated transcription coactivator 2, CRTC2, TORC-2, TORC2transducer of regulated cAMP response element-binding protein (CREB) 2, Transducer of CREB protein 2, Transducer of regulated cAMP response element-binding protein 2
Gene Symbols CRTC2
Host Species Mouse
Immunogen E. coli-derived recombinant human TORC2 Lys453-Ser613 Accession # Q53ET0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200186
Test Specificity Detects human and mouse TORC2 in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.