Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse TREM2 Alexa Fluor™ 700-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB17291N
This item is not returnable.
View return policy
Description
TREM2 Monoclonal specifically detects TREM2 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
TREM2 | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
PLOSL2, TREM-2, Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
54209 | |
Human, Mouse | |
Purified |
Flow Cytometry | |
237920 | |
Flow Cytometry 5 uL/10^6 cells | |
Q99NH8 | |
TREM2 | |
Mouse myeloma cell line NS0-derived recombinant mouse TREM-2 extracellular domain Accession # Q99NH8 | |
100 Tests | |
Primary | |
Detects human and mouse TREM-2 in direct ELISAs and Western blots. Stains TREM-2 transfectants but not TREM-1 transfectants. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction