Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse TREM2 Antibody, R&D Systems™

Rat Monoclonal Antibody
$214.00 - $514.50
Specifications
| Antigen | TREM2 |
|---|---|
| Clone | 237920R |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB17291RSP
![]() |
R&D Systems
MAB17291RSP |
25 μg |
Each for $214.00
|
|
|||||
MAB17291R10
![]() |
R&D Systems
MAB17291R100 |
100 μg |
Each for $514.50
|
|
|||||
Description
TREM2 Monoclonal antibody specifically detects TREM2 in Human, Mouse samples. It is validated for Flow CytometrySpecifications
| TREM2 | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| Monoclonal | |
| Rat | |
| Human, Mouse | |
| Q99NH8 | |
| 54209 | |
| IgG2b | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 237920R | |
| Flow Cytometry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| PLOSL2, TREM-2, Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse TREM2, Met1-Pro168, Accession # Q99NH8 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title