Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse TREM2 APC-conjugated Antibody, R&D Systems™
GSA_VAGREENER_CHOICESDP

Catalog No. FAB17291A Shop All Novus Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB17291A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB17291A Supplier R&D Systems Supplier No. FAB17291A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 5 publications

TREM2 Monoclonal specifically detects TREM2 in Human∣Mouse∣Transgenic Mouse samples. It is validated for Flow Cytometry∣Immunocytochemistry∣Neutralization.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Antigen TREM2
Applications Flow Cytometry, Immunocytochemistry, Neutralization
Classification Monoclonal
Clone 237920
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q99NH8
Gene Alias PLOSL2, TREM-2, Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2
Gene Symbols TREM2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse TREM-2 extracellular domain Accession # Q99NH8
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54209
Test Specificity Detects human and mouse TREM-2 in direct ELISAs and Western blots. Stains TREM-2 transfectants but not TREM-1 transfectants.
Target Species Human, Mouse, Transgenic Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Product Type Antibody
Form Purified
Isotype IgG2b
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.