Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse TREM2 PerCP-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB17291C
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB17291C 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB17291C Supplier R&D Systems Supplier No. FAB17291C
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

TREM2 Monoclonal specifically detects TREM2 in Human, Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen TREM2
Applications Flow Cytometry
Classification Monoclonal
Clone 237920
Conjugate PerCP
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q99NH8
Gene Alias PLOSL2, TREM-2, Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2
Gene Symbols TREM2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse TREM-2 extracellular domain Accession # Q99NH8
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54209
Test Specificity Detects human and mouse TREM-2 in direct ELISAs and Western blots. Stains TREM-2 transfectants but not TREM-1 transfectants.
Target Species Human, Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.