Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse TSC2 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems MAB40401
This item is not returnable.
View return policy
Description
TSC2 Monoclonal specifically detects TSC2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
TSC2 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 3-25 ug/mL | |
P49815 | |
TSC2 | |
E. coli-derived recombinant human TSC2 Val550-Leu850 Accession # P49815 | |
100 μg | |
Primary | |
Detects human and mouse TSC2 in Western blots. | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
614204 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
LAM, LAMFLJ43106, TSC4, Tuberin, tuberous sclerosis 2, Tuberous sclerosis 2 protein | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7249 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction