Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse TSG-6 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
$214.00 - $549.50
Specifications
Antigen | TSG-6 |
---|---|
Clone | 259820 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
TSG-6 Monoclonal specifically detects TSG-6 in Human, Mouse samples. It is validated for Western Blot.Specifications
TSG-6 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
P98066 | |
7130 | |
Mouse myeloma cell line NS0-derived recombinant human TSG-6 Trp18-Leu277 Accession # P98066 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human and mouse TSG-6 in direct ELISAs and Western blots. |
259820 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Hyaluronate-binding protein, TNF alpha-induced protein 6, TNFAIP6, TNF-stimulated gene 6 protein, TSG6, TSG-6tumor necrosis factor alpha-inducible protein 6, TSG6tumor necrosis factor-inducible gene 6 protein, Tumor necrosis factor alpha-induced protein 6, tumor necrosis factor, alpha-induced protein 6, tumor necrosis factor-stimulated gene-6 protein | |
TNFAIP6 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title