Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse UBE2K/E2-25K Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6609
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6609 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6609 Supplier R&D Systems Supplier No. MAB6609
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

UBE2K/E2-25K Monoclonal specifically detects UBE2K/E2-25K in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen UBE2K/E2-25K
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 701316
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P61086
Gene Alias E2(25K), E225K, E2-25K, EC 6.3.2.19, Hip2, HIP-2, huntingtin interacting protein 2, Huntingtin-interacting protein 2, HYPG, LIG, UbcH1, UBE2K, UBE2K-25K, ubiquitin carrier protein, ubiquitin-conjugating enzyme E2 K, Ubiquitin-conjugating enzyme E2(25K), Ubiquitin-conjugating enzyme E2-25 kDa, Ubiquitin-conjugating enzyme E2-25K, ubiquitin-conjugating enzyme E2K (UBC1 homolog, yeast), ubiquitin-protein ligase
Gene Symbols UBE2K
Host Species Mouse
Immunogen E. coli-derived recombinant human UBE2K/E2-25K Ala2-Asn200 Accession # P61086
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3093
Test Specificity Detects human UBE2K/E2-25K in direct ELISAs and Western blots.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.