Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse UCP1 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC6158R100UG
This item is not returnable.
View return policy
Description
UCP1 Monoclonal specifically detects UCP1 in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
UCP1 | |
Monoclonal | |
Alexa Fluor 647 | |
Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide. | |
SLC25A7, SLC25A7mitochondrial brown fat uncoupling protein 1, Solute carrier family 25 member 7, thermogenin, UCP, UCP 1, uncoupling protein 1 (mitochondrial, proton carrier) | |
E. coli-derived recombinant human UCP1 Met1-Thr307 Accession # P25874 | |
100 μg | |
7350 | |
Human, Mouse | |
Supernatant |
Flow Cytometry | |
536435 | |
Intracellular Staining by Flow Cytometry 0.25-1μg/106 cells | |
P25874 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human and mouse UCP1 in Western blots. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction