Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse UCP1 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC6158P100
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC6158P100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC6158P100 Supplier R&D Systems Supplier No. IC6158P100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

UCP1 Monoclonal specifically detects UCP1 in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen UCP1
Applications Flow Cytometry
Classification Monoclonal
Clone 536435
Conjugate R-PE
Dilution Intracellular Staining by Flow Cytometry 10 μL/106 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P25874
Gene Alias SLC25A7, SLC25A7mitochondrial brown fat uncoupling protein 1, Solute carrier family 25 member 7, thermogenin, UCP, UCP 1, uncoupling protein 1 (mitochondrial, proton carrier)
Gene Symbols UCP1
Host Species Mouse
Immunogen E. coli-derived recombinant human UCP1, Met1-Thr307, Accession No. P25874
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Primary or Secondary Primary
Gene ID (Entrez) 7350.0
Test Specificity Detects human and mouse UCP1 in Western blots.
Target Species Human, Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.