Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse UCP2 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF4739SP
This item is not returnable.
View return policy
Description
UCP2 Polyclonal specifically detects UCP2 in Human, Mouse samples. It is validated for Western Blot, Simple Western.Specifications
UCP2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
mitochondrial uncoupling protein 2, SLC25A8, SLC25A8BMIQ4, Solute carrier family 25 member 8, UCP 2, UCPH, uncoupling protein 2 (mitochondrial, proton carrier) | |
Goat | |
Affinity Purified | |
RUO | |
7351 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
P55851 | |
UCP2 | |
E. coli-derived recombinant human UCP2 Met1-Phe309 Accession # P55851 | |
25 μg | |
Primary | |
Detects human and mouse UCP2 in Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) UCP1 is observed. | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction