Learn More
Human/Mouse UCP2 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | UCP2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL, Simple Western 10 ug/mL |
| Gene Accession No. | P55851 |
| Gene Alias | mitochondrial uncoupling protein 2, SLC25A8, SLC25A8BMIQ4, Solute carrier family 25 member 8, UCP 2, UCPH, uncoupling protein 2 (mitochondrial, proton carrier) |
| Gene Symbols | UCP2 |
| Host Species | Goat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.