Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Vav-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3786
This item is not returnable.
View return policy
Description
Vav-1 Monoclonal specifically detects Vav-1 in Human, Mouse samples. It is validated for Western Blot.Specifications
Vav-1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
P15498 | |
VAV1 | |
E. coli-derived recombinant human Vav-1 Trp25-Leu228 Accession # P15498 | |
100 μg | |
Primary | |
Detects human and mouse Vav-1 in Western blots. | |
Human, Mouse | |
Purified |
Western Blot | |
591529 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
oncogene vav, proto-oncogene vav, vav 1 guanine nucleotide exchange factor, vav proto-oncogene, Vav1, vav-T, VAVvav 1 oncogene | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7409 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction