Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse VEGF-B167/186 Antibody, R&D Systems™

Catalog No. MAB751100
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB751100 100 μg
MAB751500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB751100 Supplier R&D Systems Supplier No. MAB751100
This item is not returnable. View return policy

Mouse Monoclonal Antibody

VEGF-B Monoclonal specifically detects VEGF-B in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen VEGF-B
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 58013
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 3-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAB06274
Gene Alias vascular endothelial growth factor B, VEGFB, VEGF-B, VEGF-related factor, VRFVEGFL
Gene Symbols VEGFB
Host Species Mouse
Immunogen E. coli-derived recombinant human VEGF-B 167 Pro22-Arg188 Accession # AAB06274
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7423
Test Specificity Detects human and mouse VEGF-B in direct ELISAs and Western blots. Recognizes both the VEGF-B 167 and VEGF-B 186 isoforms. In Western blots, no cross-reactivity with recombinant human (rh) VEGF, rhVEGF-C, rhVEGF-D, rhCTGF, rhPlGF, rhLDGF, or rhPDGF is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.